Merck
  • Differential fluctuation in virulence and VOC profiles among different cultures of entomopathogenic fungi.
Differential fluctuation in virulence and VOC profiles among different cultures of entomopathogenic fungi.
Journal of invertebrate pathology (2010-03-18)
Abid Hussain, Ming-Yi Tian, Yu-Rong He, Yan-Yuan Lei
ABSTRACT
Insect-passaged cultures of entomopathogenic fungi grown on potato dextrose agar media have been shown to have altered virulence and profiles of volatile compounds. The present study demonstrated the pathogenic status of FS(0) (in vitro) and FS(1) and FS(2) (insect-passaged cultures grown on PDA) cultures of Metarhizium anisopliae (strains 406 and 02049) and Beauveria bassiana by a non-choice assay, in which filter paper was inoculated with fungal spores at a concentration of 1 x 10(7) spores/ml. The FS(1) and FS(2) cultures of M. anisopliae strain 02049 and B. bassiana produced conidia with high virulence, and the volatile profiles of these conidia comprised relatively lower percentages of branched-alkanes than conidia from the FS(0) cultures. In contrast, the conidia from an FS(0) culture of M. anisopliae strain 406 had somewhat elevated virulence levels, but their volatile profile had <2% branched-alkanes. The FS(1) and FS(2) cultures of M. anisopliae strain 406 did not gain virulence, and these cultures showed a decline in virulence along with major alteration of their volatile profiles. Their volatile profiles mainly comprised branched-alkanes. The volatile profiles of the FS(1) and FS(2) cultures lacked n-tetradecane, which was an important component of all the virulent cultures. Four compounds, 2-phenylpropenal, 2,5,5-trimethyl-1-hexene, n-tetradecane and 2,6-dimethylheptadecane, were detected only from the virulent cultures, suggesting that low LT(50) values were probably due to the production of these compounds. This is the first report to characterize volatiles from FS(0), FS(1) and FS(2) cultures of entomopathogenic fungi; its utility in different aspects opens an interesting area for further investigations.
Sigma-Aldrich
Tetradecane, olefin free, ≥99.0% (GC)
Sigma-Aldrich
Tetradecane, ≥99%
Supelco
Tetradecane, analytical standard