Merck
  • NSAIDs in combination therapy for the treatment of chronic pseudophakic cystoid macular edema.
NSAIDs in combination therapy for the treatment of chronic pseudophakic cystoid macular edema.
Retina (Philadelphia, Pa.) (2010-02-23)
Keith A Warren, Hasan Bahrani, Jenni E Fox
ABSTRACT
The purpose of this study was to evaluate the addition of topical nonsteroidal antiinflammatory drugs (NSAIDs) to intravitreal corticosteroid and antivascular endothelial growth factor injections for the treatment of chronic cystoid macular edema. Thirty-nine patients with chronic pseudophakic cystoid macular edema completed a single-center, randomized, investigator-masked study. All patients were treated with an intravitreal triamcinolone and bevacizumab injection at study entry; the bevacizumab injection was repeated at 1 month. To evaluate the effect of adding an NSAID, patients were randomized to treatment with 1 of 4 topical NSAIDs (diclofenac 0.1%, ketorolac 0.4%, nepafenac 0.1%, and bromfenac 0.09%) or placebo for 16 weeks. At Weeks 12 and 16, both the nepafenac and bromfenac groups showed a significant reduction in retinal thickness compared with that in placebo (nepafenac, P = 0.0048, bromfenac, P = 0.0113). A difference, however, between these 2 NSAID groups was observed in that only the nepafenac group was able to maintain the demonstrated retinal thickness decrease at Weeks 12 and 16. The nepafenac group also experienced a significant improvement in visual acuity at Weeks 12 (P = 0.0084) and 16 (P = 0.0233). The addition of NSAIDs did not produce an increase in mean intraocular pressure over the course of therapy. Although NSAID therapy seems to potentiate the improvements produced by corticosteroids and antivascular endothelial growth factor therapy for chronic pseudophakic cystoid macular edema, only nepafenac- and bromfenac-treated eyes showed reduced retinal thickness at 12 weeks and 16 weeks. Furthermore, nepafenac produced a sustained improvement in visual acuity.
Sigma-Aldrich
Bromfenac sodium, ≥98% (HPLC)