Merck
Red wine phenolic complexes and their in vitro antioxidant activity.
Journal of agricultural and food chemistry (2009-09-01)
Baoshan Sun, Isabel Spranger, Jingyu Yang, Conceiço Leandro, Lei Guo, Sónia Canário, Yuqing Zhao, Chunfu Wu
ABSTRACT
Phenolic complexes are a major group of polyphenols in aged red wine. The objective of this work was to evaluate the in vitro antioxidant activity of the phenolic complexes. Thus, red wine polyphenols were fractionated into various fractions including monomers, oligomers, polymers, anthocyanins, and complexes. The in vitro antioxidant activities of these fractions and other phenolic standards (catechin, epicatechin, quercetin, and malvidin 3-glucoside) as well as ascorbic acid were verified by DPPH* test. On the other hand, the variation of antioxidant activities during the reaction between epicatechin and malvidin 3-glucoside mediated by acetaldehyde in a model wine solution was also monitored. The results showed that both the phenolic complex fraction and newly formed condensation products between epicatechin and malvidin 3-glucoside maintain antioxidant activities as strong as those of their compositional phenolics. This work provides, for the first time, direct evidence about the in vitro antioxidant activities of red wine phenolic complexes.
Sigma-Aldrich
Oenin chloride, ≥90% (HPLC)