Merck
  • Ferrous iron production mediated by tetrathionate hydrolase in tetrathionate-, sulfur-, and iron-grown Acidithiobacillus ferrooxidans ATCC 23270 cells.
Ferrous iron production mediated by tetrathionate hydrolase in tetrathionate-, sulfur-, and iron-grown Acidithiobacillus ferrooxidans ATCC 23270 cells.
Bioscience, biotechnology, and biochemistry (2009-06-09)
Tsuyoshi Sugio, Taher M Taha, Fumiaki Takeuchi
ABSTRACT
When tetrathionate-grown Acidithiobacillus ferrooxidans ATCC 23270 cells were incubated with ferric ions and tetrathionate at pH 3.0, ferrous ions were produced enzymatically. Fe(3+)-reductase, which catalyzes Fe(3+) reduction with tetrathionate, was purified to homogeneity not only from tetrathionate-grown, but also from sulfur- and iron-grown A. ferrooxidans ATCC 23270 cells. The results for apparent molecular weight measured by SDS-PAGE (52.3 kD) and the N-terminal amino acid sequences of the purified enzymes from iron-, sulfur, and tetrathionate-grown cells (AVAVPMDSTG) indicate that Fe(3+)-reductase corresponds to tetrathionate hydrolase. The evidence that tetrathionate-grown A. ferrooxidans ATCC 23270 cells have high iron-oxidizing activity at the early log phase, comparable to that of iron-grown ATCC 23270 cells, is supported by our finding that tetrathionate hydrolase produces Fe(2+) from tetrathionate during growth on tetrathionate. This is the first report on ferric reductase activity associated with tetrathionate hydrolase.
Sigma-Aldrich
Sodium tetrathionate dihydrate, ≥98% (titration)
Sigma-Aldrich
Sodium tetrathionate dihydrate, purum p.a., ≥98.0% (T)