Merck
  • Resistance to insect growth regulator insecticides in populations of sheep lice as assessed by a moulting disruption assay.
Resistance to insect growth regulator insecticides in populations of sheep lice as assessed by a moulting disruption assay.
Medical and veterinary entomology (2009-01-06)
P J James, A P Cramp, S E Hook
ABSTRACT
Low-volume, backline applications with the benzoylphenyl urea insecticides triflumuron and diflubenzuron represent in excess of 70% of treatments for the control of sheep lice, Bovicola ovis (Schrank) (Phthiraptera: Trichodectidae), in Australia. Reports of reduced effectiveness from 2003 and subsequent controlled treatment trials suggested the emergence of resistance to these compounds in B. ovis populations. A laboratory assay based on the measurement of moulting success in nymphs was developed and used to assess susceptibility to diflubenzuron and triflumuron in louse populations collected from sheep where a control failure had occurred. These tests confirmed the development of resistance to triflumuron and diflubenzuron in at least two instances, with estimated resistance ratios of 67-94X at LC(50).
Product Description
Supelco
Triflumuron, PESTANAL®, analytical standard