Merck
  • Metabolism of hydroxylated and fluorinated benzoates by Syntrophus aciditrophicus and detection of a fluorodiene metabolite.
Metabolism of hydroxylated and fluorinated benzoates by Syntrophus aciditrophicus and detection of a fluorodiene metabolite.
Applied and environmental microbiology (2008-12-31)
Housna Mouttaki, Mark A Nanny, Michael J McInerney
ABSTRACT
Transformations of 2-hydroxybenzoate and fluorobenzoate isomers were investigated in the strictly anaerobic Syntrophus aciditrophicus to gain insight into the initial steps of the metabolism of aromatic acids. 2-Hydroxybenzoate was metabolized to methane and acetate by S. aciditrophicus and Methanospirillum hungatei cocultures and reduced to cyclohexane carboxylate by pure cultures of S. aciditrophicus when grown in the presence of crotonate. Under both conditions, transient accumulation of benzoate but not phenol was observed, indicating that dehydroxylation occurred prior to ring reduction. Pure cultures of S. aciditrophicus reductively dehalogenated 3-fluorobenzoate with the stoichiometric accumulation of benzoate and fluorine. 3-Fluorobenzoate-degrading cultures produced a metabolite that had a fragmentation pattern almost identical to that of the trimethylsilyl (TMS) derivative of 3-fluorobenzoate but with a mass increase of 2 units. When cells were incubated with deuterated water, this metabolite had a mass increase of 3 or 4 units relative to the TMS derivative of 3-fluorobenzoate. (19)F nuclear magnetic resonance spectroscopy ((19)F NMR) detected a metabolite in fluorobenzoate-degrading cultures with two double bonds, either 1-carboxyl-3-fluoro-2,6-cyclohexadiene or 1-carboxyl-3-fluoro-3,6-cyclohexadiene. The mass spectral and NMR data are consistent with the addition of two hydrogen or deuterium atoms to 3-fluorobenzoate, forming a 3-fluorocyclohexadiene metabolite. The production of a diene metabolite provides evidence that S. aciditrophicus contains dearomatizing reductase that uses two electrons to dearomatize the aromatic ring.
Sigma-Aldrich
Cyclohexanecarboxylic acid, 98%
Sigma-Aldrich
3-Fluorobenzoic acid, 97%