Merck
Fragrance material review on fenchyl alcohol.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2008-07-22)
S P Bhatia, D McGinty, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review of fenchyl alcohol when used as a fragrance ingredient is presented.
Sigma-Aldrich
Fenchyl alcohol, ≥96%, FG