Merck
  • Towards 'bio-based' Nylon: conversion of gamma-valerolactone to methyl pentenoate under catalytic distillation conditions.
Towards 'bio-based' Nylon: conversion of gamma-valerolactone to methyl pentenoate under catalytic distillation conditions.
Chemical communications (Cambridge, England) (2007-08-19)
Jean-Paul Lange, Jan Z Vestering, René J Haan
ABSTRACT
Methyl pentenoate, a promising Nylon intermediate, is produced in >95% yield via the transesterification of gamma-valerolactone, a bio-based intermediate, under catalytic distillation conditions.
Sigma-Aldrich
γ-Valerolactone, ≥99%, FCC, FG
Sigma-Aldrich
γ-Valerolactone, ReagentPlus®, 99%
Sigma-Aldrich
γ-Valerolactone, natural, ≥95%, FG