Merck
  • Quantification of active principles and pigments in leaf extracts of Isatis tinctoria by HPLC/UV/MS.
Quantification of active principles and pigments in leaf extracts of Isatis tinctoria by HPLC/UV/MS.
Planta medica (2007-01-20)
Tobias Mohn, Olivier Potterat, Matthias Hamburger
ABSTRACT
An HPLC method has been developed and validated for the quantification of the pharmacologically active principles tryptanthrin (1), 1,3-dihydro-3-[(4-hydroxy-3,5-dimethoxyphenyl)methylene]-2 H-indol-2-one (indolinone) (3), indirubin (4), alpha-linolenic acid (2), and indigo (5), an isomer of indirubin, in extracts from the traditional anti-inflammatory plant Isatis tinctoria (woad). The chromatographic separation was performed on a C-18 column with a linear gradient of acetonitrile in water containing 0.1% formic acid. The method combines UV and electrospray MS detection in the positive ion mode for the detection of the alkaloids, with a switch to the negative mode for the analysis of alpha-linolenic acid. The method was applied to the analysis of woad extracts obtained by supercritical fluid (SFE) CO2 extraction, and by pressurized liquid extraction (PLE) with dichloromethane and methanol, respectively. While the highest concentration of alpha-linolenic acid was found in the SFE extract (7.43%), the concentrations of tryptanthrin , indolinone, indirubin and indigo were the highest in the dichloromethane extract (0.30, 0.035, 2.48 and 0.84%, respectively). Compound 3 was not detected in the methanolic extract and only traces of compounds 1, 4 and 5 and low amount of alpha-linolenic acid (0.39%) were present in this extract.
Sigma-Aldrich
Tryptanthrin, ≥98% (HPLC)