Merck
  • L-cyst(e)ine requirements of malignant cells and progress toward depletion therapy.
L-cyst(e)ine requirements of malignant cells and progress toward depletion therapy.
Cancer treatment reports (1979-06-01)
J R Uren, H Lazarus
PMID466647
ABSTRACT
The L-cyst(e)ine requirements of normal and malignant cells are reviewed and expanded within the context of establishing whether the measurement of gamma-cystathionase levels constitutes a predictive test for tumor sensitivity to L-cyst(e)ine depletion. The ability of both purified L-cysteine desulfhydrase and gamma-cystathionase to inhibit the growth of the L-cystine-dependent L1210 leukemia in culture is presented, as well as approaches to circumvent the limitations of these enzymes for in vivo therapy. The ability of proparagylglycine to inhibit L-cysteine biosynthesis in vivo is reviewed for its possible use in combination therapy. In addition, the ability of poly D,L-alanine modification of Escherichia coli L-asparaginase to increase the plasma half-life in mice tenfold as well as to decrease the immunogenicity of the enzyme is presented.