Merck
  • Substitution of glutamic acid 109 by aspartic acid alters the substrate specificity and catalytic activity of the beta-subunit in the tryptophan synthase bienzyme complex from Salmonella typhimurium.
Substitution of glutamic acid 109 by aspartic acid alters the substrate specificity and catalytic activity of the beta-subunit in the tryptophan synthase bienzyme complex from Salmonella typhimurium.
Biochemistry (1992-02-04)
P S Brzović, A M Kayastha, E W Miles, M F Dunn
ABSTRACT
In an effort to understand the catalytic mechanism of the tryptophan synthase beta-subunit from Salmonella typhimurium, possible functional active site residues have been identified (on the basis of the 3-D crystal structure of the bienzyme complex) and targeted for analysis utilizing site-directed mutagenesis. The chromophoric properties of the pyridoxal 5'-phosphate cofactor provide a particularly convenient and sensitive spectral probe to directly investigate changes in catalytic events which occur upon modification of the beta-subunit. Substitution of Asp for Glu 109 in the beta-subunit was found to alter both the catalytic activity and the substrate specificity of the beta-reaction. Steady-state kinetic data reveal that the beta-reaction catalyzed by the beta E109D alpha 2 beta 2 mutant enzyme complex is reduced 27-fold compared to the wild-type enzyme. Rapid-scanning stopped-flow (RSSF) UV-visible spectroscopy shows that the mutation does not seriously affect the pre-steady-state reaction of the beta E109D mutant with L-serine to form the alpha-aminoacrylate intermediate, E(A-A). Binding of the alpha-subunit specific ligand, alpha-glycerol phosphate (GP) to the alpha 2 beta 2 complex exerts the same allosteric effects on the beta-subunit as observed with the wild-type enzyme. However, the pre-steady-state spectral changes for the reaction of indole with E(A-A) show that the formation of the L-tryptophan quinonoid, E(Q3), is drastically altered. Discrimination against E(Q3) formation is also observed for the binding of L-tryptophan to the mutant alpha 2 beta 2 complex in the reverse reaction. In contrast, substitution of Asp for Glu 109 increases the apparent affinity of the beta E109D alpha-aminoacrylate complex for the indole analogue indoline and results in the increased rate of synthesis of the amino acid product dihydroiso-L-tryptophan. Thus, the mutation affects the covalent bond forming addition reactions and the nucleophile specificity of the beta-reaction catalyzed by the bienzyme complex.
Sigma-Aldrich
N-Methylhydroxylamine hydrochloride, 98%