Merck
  • Stereochemical assignment of naturally occurring 2,3-epoxy-2-methylbutanoate esters.
Stereochemical assignment of naturally occurring 2,3-epoxy-2-methylbutanoate esters.
Phytochemical analysis : PCA (2002-12-24)
J Martín Torres-Valencia, Guadalupe I León, J Roberto Villagómez-Ibarra, Oscar R Suárez-Castillo, Carlos M Cerda-García-Rojas, Pedro Joseph-Nathan
ABSTRACT
A method to determine the absolute configuration of 2,3-epoxy-2-methylbutanoate ester residues in natural products is presented, based on (i) the reduction of the ester function to yield a 2-methyl-1,2-butanediol, (ii) esterification of the obtained primary alcohol with either (R)-(+)- or (S)-(-)-Mosher's acid to afford the corresponding Mosher's ester, and (iii) 1H-NMR spectral comparison of the final product with that of the Mosher's esters prepared from 2-methyl-1,2-butanediols of known stereochemistry.
Sigma-Aldrich
(R)-(+)-α-Methoxy-α-trifluoromethylphenylacetic acid, 99%
Sigma-Aldrich
(S)-(−)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid, ≥99%