Merck
  • Biotransformations catalyzed by cloned p-cymene monooxygenase from Pseudomonas putida F1.
Biotransformations catalyzed by cloned p-cymene monooxygenase from Pseudomonas putida F1.
Applied microbiology and biotechnology (2001-05-09)
T Nishio, A Patel, Y Wang, P C Lau
ABSTRACT
p-Cymene monooxygenase (CMO) from Pseudomonas putida F1 consists of a hydroxylase (CymA1) and a reductase component (CymA2) which initiate pcymene (p-isopropyltoluene) catabolism by oxidation of the methyl group to p-isopropylbenzyl alcohol (p-cumic alcohol). To study the possible diverse range of substrates catalyzed by CMO, the cymA1A2 genes were cloned in an Escherichia coli pT7-5 expression system and the cells were used in transformation experiments. The tested substrates include different substituents on the aromatic ring at the 2 (ortho), 3 (meta) or 4 (para) position relative to the methyl moiety. As a result, a distinct preference was observed for substrates containing at least an alkyl or heteroatom substituent at the para-position of toluene. The conversion rate of 4-chlorotoluene or 4-methylthiotoluene to the corresponding benzyl alcohol was found to be as good as the canonical substrate, p-cymene. But 3-chlorotoluene, 4-fluorotoluene and 4-nitrotoluene were relatively poor substrates. CMO is also capable of producing styrene oxide from styrene. However, the oxidation of 4-chlorostyrene to 4-chlorostyrene oxide was by far the fastest among the substrates used in this study. The various biotransformation products were identified by a combined solid phase microextraction/gas chromatographic-mass spectrometric analytical technique.
Sigma-Aldrich
4-Chlorotoluene, 98%