Merck
  • Subcellular localization of two neurotropic drugs in three varieties of central nervous system cells by secondary ion mass spectroscopy (SIMS) microscopy.
Subcellular localization of two neurotropic drugs in three varieties of central nervous system cells by secondary ion mass spectroscopy (SIMS) microscopy.
Biology of the cell (1992-01-01)
L L Zhang, M Tardy, J P Berry, F Escaig, P Galle
ABSTRACT
The intracellular localization of two neurotropic drugs, flunitrazepam (benzodiazepine) and triflupromazine (phenothiazine), was studied by secondary ion mass spectrometry microscopy (SIMS) in three varieties of cells. The images of the intracellular distributions of the two drugs are easily obtained by selecting the fluorine atom of the molecules. These images show that the drug from the benzodiazepine group is mainly located in the nuclei, whereas the phenothiazine is exclusively located inside the cytoplasm.