Merck
[Photo-induced conformational motility of proteins].
Biofizika (2012-11-10)
N L Vekshin
ABSTRACT
The dynamics of proteins, detected by fluorescence, consists of three components: spontaneous dynamics, dipole-dipole photo-induced dynamics, thermal photo-induced dynamics. The photo-induced dynamics can lead to activation as well as inactivation of enzymes.
Sigma-Aldrich
Alcohol Dehydrogenase equine, recombinant, expressed in E. coli, ≥0.5 U/mg
Sigma-Aldrich
Alcohol Dehydrogenase from Saccharomyces cerevisiae
Sigma-Aldrich
Alcohol Dehydrogenase from Saccharomyces cerevisiae, ≥300 units/mg protein, lyophilized powder (contains buffer salts), Mw 141-151 kDa
Sigma-Aldrich
Alcohol Dehydrogenase from Saccharomyces cerevisiae, powder, ≥300 units/mg protein, mol wt ~141,000 (four subunits)