Merck
  • Topology of prostaglandin H synthase-1 in the endoplasmic reticulum membrane.
Topology of prostaglandin H synthase-1 in the endoplasmic reticulum membrane.
Archives of biochemistry and biophysics (1995-10-20)
Y Ren, C Walker, D S Loose-Mitchell, J Deng, K H Ruan, R J Kulmacz
ABSTRACT
Prostaglandin H synthase-1 is an integral endoplasmic reticulum membrane protein which catalyzes a key control step in prostaglandin biosynthesis. The overall arrangement of the prostaglandin H synthase-1 polypeptide with respect to the endoplasmic reticulum membrane was examined in transiently transfected COS-1 cells, using immunofluorescence microscopy. A bacterial toxin, streptolysin-O, was used for selective plasma membrane permeabilization and a detergent, saponin, for general membrane permeabilization. Treated cells were probed with six antibodies specific for particular prostaglandin H synthase-1 peptide segments and one antibody specific for an inserted viral reporter epitope. Control experiments established that actin, a cytoplasmic marker, was accessible to fluorescein-labeled phalloidin after streptolysin-O treatment, whereas antibodies against protein disulfide isomerase, an endoplasmic reticulum lumenal marker, bound only after saponin treatment, Using this approach to investigate prostaglandin H synthase-1, it was found that streptolysin-O treatment was sufficient to obtain staining of intracellular membranes by antibodies specific for the endogenous C-terminal segment, for the viral reporter inserted at the C-terminus, and for the protease-sensitive region near arg277. In contrast, saponin treatment was necessary for staining by antibodies specific for peptides spanning residues 51-66, 156-170, and 377-390. Antibodies targeted against residues 483-496 did not stain transfected cells even after saponin permeabilization, although they did bind to detergent-solubilized prostaglandin H synthase-1. These results indicate that the C-terminus and arg277 regions of the synthase can be exposed on the cytoplasmic side of the endoplasmic reticulum membrane, whereas regions near N-glycosylation sites are confined to the endoplasmic reticulum lumen and residues 483-496 are inaccessible from either side of the endoplasmic reticulum membrane.
Sigma-Aldrich
Phalloidin–Atto 390, BioReagent, suitable for fluorescence, ≥90% (HPLC)