Merck
Thiol-sensitive genes of Escherichia coli.
Journal of bacteriology (1989-10-01)
G T Javor
ABSTRACT
The effect of 1-thioglycerol on the expression of genes of Escherichia coli was investigated. Pulse-labeled proteins from aerobically growing, 1-thioglycerol-treated E. coli were separated by two-dimensional gel electrophoresis, and their radioactivities were compared with those of identical proteins from nontreated cells. The first 10 min of exposure to thiol stimulated the synthesis of 10% of the observed proteins and inhibited the production of 16% of the proteins. After 30 min of growth with thiol, the synthesis of 44% of the observed proteins was inhibited and synthesis of 18% of the proteins was stimulated. In general, the expression of genes of carbohydrate metabolism, amino acid metabolism, and protein biosynthesis were inhibited, while nucleic acid synthetic and repair gene expressions showed mixed responses. Synthesis of transport proteins was not affected. Transient stimulation of oxidative-stress proteins and sustained stimulation of the expressions of trxB, ompA, and ompB genes and those of several unidentified gene products were also observed. Whether these complex responses merely reflect adjustments by cellular subsystems to a suddenly reducing environment or whether they are manifestations of a reductive-stress regulon will have to await genetic analysis of this phenomenon.
Sigma-Aldrich
1-Thioglycerol, BioXtra, ≥97%