Merck
  • ThPOK is a critical multifaceted regulator of myeloid lineage development.
ThPOK is a critical multifaceted regulator of myeloid lineage development.
Nature immunology (2023-07-21)
Jayati Basu, Andre Olsson, Kyle Ferchen, Elizaveta K Titerina, Kashish Chetal, Emmanuelle Nicolas, Philip Czyzewicz, Dmitry Levchenko, Lu Ge, Xiang Hua, H Leighton Grimes, Nathan Salomonis, Dietmar J Kappes
ABSTRACT
The transcription factor ThPOK (encoded by Zbtb7b) is well known for its role as a master regulator of CD4 lineage commitment in the thymus. Here, we report an unexpected and critical role of ThPOK as a multifaceted regulator of myeloid lineage commitment, differentiation and maturation. Using reporter and knockout mouse models combined with single-cell RNA-sequencing, progenitor transfer and colony assays, we show that ThPOK controls monocyte-dendritic cell versus granulocyte lineage production during homeostatic differentiation, and serves as a brake for neutrophil maturation in granulocyte lineage-specified cells through transcriptional regulation of lineage-specific transcription factors and RNA via altered messenger RNA splicing to reprogram intron retention.
Sigma-Aldrich
Anti-IFIT1/p56 Antibody, serum, from rabbit