Merck
  • State of the Art in the Culture of the Human Microbiota: New Interests and Strategies.
State of the Art in the Culture of the Human Microbiota: New Interests and Strategies.
Clinical microbiology reviews (2020-10-30)
Maryam Tidjani Alou, Sabrina Naud, Saber Khelaifia, Marion Bonnet, Jean-Christophe Lagier, Didier Raoult
ABSTRACT
The last 5 years have seen a turning point in the study of the gut microbiota with a rebirth of culture-dependent approaches to study the gut microbiota. High-throughput methods have been developed to study bacterial diversity with culture conditions aimed at mimicking the gut environment by using rich media such as YCFA (yeast extract, casein hydrolysate, fatty acids) and Gifu anaerobic medium in an anaerobic workstation, as well as media enriched with rumen and blood and coculture, to mimic the symbiosis of the gut microbiota. Other culture conditions target phenotypic and metabolic features of bacterial species to facilitate their isolation. Preexisting technologies such as next-generation sequencing and flow cytometry have also been utilized to develop innovative methods to isolate previously uncultured bacteria or explore viability in samples of interest. These techniques have been applied to isolate CPR (Candidate Phyla Radiation) among other, more classic approaches. Methanogenic archaeal and fungal cultures present different challenges than bacterial cultures. Efforts to improve the available systems to grow archaea have been successful through coculture systems. For fungi that are more easily isolated from the human microbiota, the challenge resides in the identification of the isolates, which has been approached by applying matrix-assisted laser desorption ionization-time of flight mass spectrometry technology to fungi. Bacteriotherapy represents a nonnegligible avenue in the future of medicine to correct dysbiosis and improve health or response to therapy. Although great strides have been achieved in the last 5 years, efforts in bacterial culture need to be sustained to continue deciphering the dark matter of metagenomics, particularly CPR, and extend these methods to archaea and fungi.
Sigma-Aldrich
MediaBoost, non-animal sterile microbial growth supplement