Merck
  • IGFBP2 Drives Regulatory T Cell Differentiation through STAT3/IDO Signaling Pathway in Pancreatic Cancer.
IGFBP2 Drives Regulatory T Cell Differentiation through STAT3/IDO Signaling Pathway in Pancreatic Cancer.
Journal of personalized medicine (2022-12-24)
Longhao Sun, Yang Zhang, Tiantian Yang, Junhang Chen, Xuebin Zhang, Xiaoyu Liang
ABSTRACT
Pancreatic ductal adenocarcinoma (PDAC) represents one of the deadliest malignancies. Elevated regulatory T cell (Treg) infiltration has a potent immunosuppressive function in tumor biology, which contributes to low survival in PDAC. Nonetheless, the crosstalk between malignant cells and tumor-infiltrating Tregs in PDAC is not well understood. Here, clinical data demonstrates that the insulin-like growth factor binding protein 2 (IGFBP2) is associated with Treg accumulation in the microenvironment of PDAC in humans. Additionally, IGFBP2 increases Treg infiltration in the tumor microenvironment and promotes disease progression in mouse PDAC. Bioinformatic analysis and mechanistic assessment reveals IGFBP2 upregulated indoleamine 2, 3-dioxygenase (IDO) by activating signal transducer and activator of transcription 3 (STAT3) signaling in PDAC cells, thus inducing Treg differentiation and an immunosuppressive tumor microenvironment. These findings provide mechanistic insights into an important molecular pathway that promotes an immunosuppressive microenvironment, which suggests the IGFBP2 axis as a potential target for improved immune response in PDAC.
Sigma-Aldrich
1-Methyl-D-tryptophan, 95%