Merck
  • Inhibition of BRD4 enhanced the tumor suppression effect of dasatinib in gastric cancer.
Inhibition of BRD4 enhanced the tumor suppression effect of dasatinib in gastric cancer.
Medical oncology (Northwood, London, England) (2022-11-10)
Hao Shen, Xuefei Hu, Xinrui Yang, Jiahui Chen, Yating Fu, Hongwei He, Yongkang Shi, Rong Zeng, Wenjun Chang, Shangyong Zheng
ABSTRACT
BRD4, a member of the bromodomain and extraterminal (BET) family, is elevated in multiple cancer tissues, including gastric cancer (GC). Targeted therapy with BRD4 may help improve the overall survival of patients with GC. Meanwhile, the approved multi-target kinase inhibitor, dasatinib, was recently reported to show varied tumor-suppressive effects in GC cells. This study investigated BRD4 expression in vivo and in vitro using immunohistochemistry and western blotting, respectively. We discussed the relationship between BRD4 expression and patient prognosis. Next, the antitumor efficacy of dasatinib was measured in BRD4-knockdown GC cells to determine the role of BRD4 blockage in dasatinib treatment. Finally, molibresib, a BET inhibitor, was used to measure the cooperative function of BRD4 inhibition and dasatinib treatment in three GC cell lines. Epithelial BRD4 expression was higher in tumoral and metastatic tissues and was strongly associated with unfavorable tumor, node, and metastasis stages and survival. BRD4 expression was heterogeneous in the three GC cell lines tested in vitro. In SGC7901, a BRD4-high GC cell line, knockdown of BRD4 using specific siRNAs suppressed cell growth individually and cooperatively with dasatinib. Moreover, molibresib and dasatinib showed a cooperative effect in suppressing the proliferation of BRD4-high GC cells. In conclusion, we confirmed that increased epithelial BRD4 expression is associated with poor disease stage and prognosis in GC and BRD4 blockage might be a valuable strategy to improve the sensitivity of dasatinib and other drugs in the chemotherapy of advanced GC.
Sigma-Aldrich
Anti-NNMT antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution