Merck
  • Histological and lectin histochemical studies of the vomeronasal organ of horses.
Histological and lectin histochemical studies of the vomeronasal organ of horses.
Tissue & cell (2016-05-29)
Kwang-Hyup Lee, Changnam Park, Jeongtae Kim, Changjong Moon, Meejung Ahn, Taekyun Shin
ABSTRACT
The morphological characteristics and glycoconjugate composition of the vomeronasal organ (VNO) of the horse was investigated using histological, immunohistochemical, and lectin histochemical methods. The VNO is bilaterally located at the base of the nasal septum, has a tubular structure surrounded by cartilage, and consists of sensory and non-sensory epithelia. Immunohistochemical examination showed that the vomeronasal sensory epithelium (VSE) consisted of receptor cells positive for both olfactory marker protein (OMP) and protein gene product 9.5 (PGP 9.5), supporting cells, and basal cells. VNO receptor cells were positive for G protein Gαi2 (vomeronasal receptor type 1 marker), but not Gαo (vomeronasal receptor type 2 marker). Lectin histochemical studies using 21 biotinylated lectins showed that the free border of the VSE was positive for 20 lectins. The receptor and supporting cells reacted with 16 lectins while the basal cells reacted with 15 lectins, with varying intensities. In the vomeronasal non-sensory epithelium, the free border was positive for 19 lectins. The cilated cells were positive for 17 lectins and the basal cells were positive for 15 lectins. The vomeronasal glands, positioned in the lamina propria, were stained with both periodic acid Schiff (PAS) and alcian blue (pH 2.5). Eighteen lectins stained the acinar cells of the vomeronasal glands with various binding patterns. These findings suggest that horse VNO receptor cells express vomeronasal receptor type 1, and the VNO glands have mucous to seromucous characteristics. Moreover, each lectin differentially binds each cell type in both the VNO sensory and non-sensory epithelia.
Sigma-Aldrich
β-Galactose Dehydrogenase from Pseudomonas fluorescens, recombinant, expressed in E. coli, ammonium sulfate suspension, ≥50 units/mg protein (biuret)
Sigma-Aldrich
N-Acetyl-D-glucosamine, ≥99% (HPLC)