Merck
  • Indigo plant leaf extract inhibits the binding of SARS-CoV-2 spike protein to angiotensin-converting enzyme 2.
Indigo plant leaf extract inhibits the binding of SARS-CoV-2 spike protein to angiotensin-converting enzyme 2.
Experimental and therapeutic medicine (2022-03-08)
Man Hagiyama, Fuka Takeuchi, Aki Sugano, Azusa Yoneshige, Takao Inoue, Akihiro Wada, Hiroshi Kajiyama, Yutaka Takaoka, Kenroh Sasaki, Akihiko Ito
ABSTRACT
Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) uses its S1 spike protein to bind to angiotensin-converting enzyme 2 (ACE2) on human cells in the first step of cell entry. Tryptanthrin, extracted from leaves of the indigo plant, Polygonum tinctorium, using d-limonene (17.3 µg/ml), is considered to inhibit ACE2-mediated cell entry of another type of coronavirus, HCoV-NL63. The current study examined whether this extract could inhibit the binding of the SARS-CoV-2 spike protein to ACE2. Binding was quantified as cell-bound fluorescence intensity in live cell cultures in which canine kidney MDCK cells overexpressing ACE2 were incubated with fluorescein-labeled S1 spike protein. When indigo extract, together with S1 protein, was added at 8,650x and 17,300x dilutions, fluorescence intensity decreased in a dose- and S1 extract-dependent manner, without affecting cell viability. When 4.0-nM tryptanthrin was added instead of the indigo extract, fluorescence intensity also decreased, but to a lesser degree than with indigo extract. Docking simulation analyses revealed that tryptanthrin readily bound to the receptor-binding domain of the S1 protein, and identified 2- and 7-amino acid sequences as the preferred binding sites. The indigo extract appeared to inhibit S1-ACE2 binding at high dilutions, and evidently contained other inhibitory elements as well as tryptanthrin. This extract may be useful for the prevention or treatment of SARS-CoV-2 infection.
Sigma-Aldrich
Tryptanthrin, ≥98% (HPLC)