Merck
  • Fluorescence diagnosis of metastatic lymph nodes using 5-aminolevulinic acid (5-ALA) in a mouse model of colon cancer.
Fluorescence diagnosis of metastatic lymph nodes using 5-aminolevulinic acid (5-ALA) in a mouse model of colon cancer.
The Journal of surgical research (2012-01-10)
Shigeru Kato, Junichiro Kawamura, Kenji Kawada, Suguru Hasegawa, Yoshiharu Sakai
ABSTRACT
Lymph node metastasis is one of the most critical prognostic factors in patients with colorectal cancer. Although regional lymph nodes should be surgically resected and pathologically examined, techniques for the intraoperative diagnosis of lymph node metastasis remain to be well established. Fluorescence diagnosis using 5-aminolevulinic acid (5-ALA) is a promising technique for evaluating various malignancies. After exogenous administration of 5-ALA, protoporphyrin IX (PPIX) accumulates in malignant cells and can be detected as red fluorescence. In this study, we investigated the usefulness of fluorescence diagnosis using 5-ALA for the detection of lymph node metastasis in a mouse model of colon cancer. An orthotopic colon cancer model was prepared by inoculating the cecal wall of nude mice with HCA7, a human colon adenocarcinoma cell line. After 3 wk, 40 mg/kg of 5-ALA was administered intraperitoneally (IP) or orally (PO). Fluorescence diagnosis with a D-Light System (Karl Storz) was then performed after 3 or 6 h. In the IP group, PPIX fluorescence was detected in metastatic lymph nodes as well as in other malignant lesions, including primary tumors and abdominal implantations, while non-metastatic nodes were fluorescence-negative. In contrast, no obvious fluorescence was detected in cancerous tissues in the PO group. PPIX fluorescence induced by intraperitoneal injection of 5-ALA allows metastatic lymph nodes to be accurately diagnosed in this mouse model. This technique may facilitate the intraoperative diagnosis of lymph node metastases from colon cancer in a clinical setting.
Sigma-Aldrich
5-Aminolevulinic acid hydrochloride, ≥98%