Merck
  • Anticancer roles of let-7f-1-3p in non-small cell lung cancer via direct targeting of integrin β1.
Anticancer roles of let-7f-1-3p in non-small cell lung cancer via direct targeting of integrin β1.
Experimental and therapeutic medicine (2021-10-12)
Yanan Yang, Yuanrong Liu, Ning Xie, Liying Shao, Hang Sun, Yubo Wei, Yunxiao Sun, Pingyu Wang, Yunfei Yan, Shuyang Xie, Youjie Li
ABSTRACT
Lung cancer is one of the most common types of cancer, with the highest mortality rate worldwide. MicroRNAs play notable roles in the chemotherapeutic effects of anticancer drugs. The present study used reverse transcription-quantitative PCR, western blotting and cell migration and invasion assays to reveal the role of let-7f-1-3p in non-small cell lung cancer (NSCLC) and explore the effect of let-7f-1-3p on doxorubicin (DOX) treatment. It was demonstrated that the levels of let-7f-1-3p in carcinoma tissues were lower compared with those in paracarcinoma tissues. Thus, let-7f-1-3p may act as a suppressor gene. The present study also explored the role of let-7f-1-3p in A549 and NCI-H1975 cells. Results revealed that let-7f-1-3p could inhibit the viability, migration and invasion of NSCLC cells and induce their apoptosis. Integrin β1 acted as a target gene regulated by let-7f-1-3p. This suggested that let-7f-1-3p could enhance DOX-inhibited cell viability, migration and invasion in vitro. Overall, the present study demonstrated that let-7f-1-3p may act as a target for drug design and lung cancer therapy.
Product Description
Roche
cOmplete Protease Inhibitor Cocktail, Tablets provided in glass vials