- Binding stoichiometry of a fluorescent cGMP analogue to membranes of retinal rod outer segments.
Binding stoichiometry of a fluorescent cGMP analogue to membranes of retinal rod outer segments.
European journal of biochemistry (1985-11-15)
A Caretta, A Cavaggioni, R T Sorbi
PMID2998793
ABSTRACT
The high-affinity binding of the cGMP analogue 8-(5-thioacetamidofluorescein)-cGMP to rod outer segment membranes depleted of peripherally bound proteins has been defined by equilibrium dialysis (mean +/- SD): membranes contain about one cGMP binding site per 130 rhodopsin molecules; the concentration of free ligand for half saturation is 2.0 +/- 0.6 microM; the apparent Hill coefficient of the bound versus free ligand relationship is 1.7 +/- 0.5; half saturation of the binding sites is sufficient for 85% activation of calcium permeability. A gating mechanism is proposed.