Merck
  • Characterization of parathyroid hormone/parathyroid hormone-related protein receptor and signaling in hypercalcemic Walker 256 tumor cells.
Characterization of parathyroid hormone/parathyroid hormone-related protein receptor and signaling in hypercalcemic Walker 256 tumor cells.
The Journal of endocrinology (2000-06-17)
P Esbrit, J Benítez-Verguizas, F de Miguel, A Valín, A García-Ocaña
ABSTRACT
Parathyroid hormone (PTH)-related protein (PTHrP) is the main factor responsible for humoral hypercalcemia of malignancy. Both PTH and PTHrP bind to the common type I PTH/PTHrP receptor (PTHR), thereby activating phospholipase C and adenylate cyclase through various G proteins, in bone and renal cells. However, various normal and transformed cell types, including hypercalcemic Walker 256 (W256) tumor cells, do not produce cAMP after PTHrP stimulation. We characterized the PTHrP receptor and the signaling mechanism upon its activation in the latter cells. Scatchard analysis of PTHrP-binding data in W256 tumor cells revealed the presence of high affinity binding sites with an apparent K(d) of 17 nM, and a density of 90 000 sites/cell. In addition, W256 tumor cells immunostained with an anti-PTHR antibody, recognizing its extracellular domain. Furthermore, reverse transcription followed by PCR, using primers amplifying two different regions in the PTHR cDNA corresponding to the N- and C-terminal domains, yielded products from W256 tumor cell RNA which were identical to the corresponding products obtained from rat kidney RNA. Consistent with our previous findings on cAMP production, 1 microM PTHrP(1-34), in contrast to 10 microg/ml cholera toxin or 1 microM isoproterenol, failed to affect protein kinase A activity in W256 tumor cells. However, in these cells we found a functional PTHR coupling to G(alpha)(q/11), whose presence was demonstrated in these tumor cell membranes by Western blot analysis. Our findings indicate that W256 tumor cells express the PTHR, which seems to be coupled to G(alpha)(q/11). Taken together with previous data, these results support the hypothesis that a switch from the cAMP pathway to the phospholipase C-intracellular calcium pathway, associated with PTHR activation, occurs in malignant cells.
Sigma-Aldrich
Hypercalcemia of malignancy factor fragment 1-34 amide human, ≥97% (HPLC)
Sigma-Aldrich
Hypercalcemia of malignancy factor 1-40 human, ≥95% (HPLC)