MilliporeSigma
  • The mitochondrial citrate transporter, CIC, is essential for mitochondrial homeostasis.
The mitochondrial citrate transporter, CIC, is essential for mitochondrial homeostasis.
Oncotarget (2012-10-27)
Olga Catalina-Rodriguez, Vamsi K Kolukula, York Tomita, Anju Preet, Ferdinando Palmieri, Anton Wellstein, Stephen Byers, Amato J Giaccia, Eric Glasgow, Chris Albanese, Maria Laura Avantaggiati
ABSTRACT
Dysregulation of the pathways that preserve mitochondrial integrity hallmarks many human diseases including diabetes, neurodegeration, aging and cancer. The mitochondrial citrate transporter gene, SLC25A1 or CIC, maps on chromosome 22q11.21, a region amplified in some tumors and deleted in developmental disorders known as velo-cardio-facial- and DiGeorge syndromes. We report here that in tumor cells CIC maintains mitochondrial integrity and bioenergetics, protects from mitochondrial damage and circumvents mitochondrial depletion via autophagy, hence promoting proliferation. CIC levels are increased in human cancers and its inhibition has anti-tumor activity, albeit with no toxicity on adult normal tissues. The knock-down of the CIC gene in zebrafish leads to mitochondria depletion and to proliferation defects that recapitulate features of human velo-cardio-facial syndrome, a phenotype rescued by blocking autophagy. Our findings reveal that CIC maintains mitochondrial homeostasis in metabolically active, high proliferating tissues and imply that this protein is a therapeutic target in cancer and likely, in other human diseases.