Merck
  • Ultracentrifugal separation, characterization, and functional study of extracellular vesicles derived from serum-free cell culture.
Ultracentrifugal separation, characterization, and functional study of extracellular vesicles derived from serum-free cell culture.
STAR protocols (2021-07-06)
Hooi Ting Hu, Tamako Nishimura, Shiro Suetsugu
ABSTRACT
Extracellular vesicles (EVs) play important roles in extracellular trafficking and signaling. Here, we separate EVs by differential centrifugation. EVs separated by this approach are called large EVs (l-EVs) and small EVs (s-EVs), reflecting particle size, which sediment based on different ultracentrifugation forces. The resulting EVs can be quantified and analyzed using nanoparticle tracking analysis, immunoblotting, and functional assays. This protocol was applied to a suspension cell line with high transfection efficiency adapted to a high-density, serum-free culture. For complete details on the use and execution of this protocol, please refer to Nishimura et al. (2021).
Sigma-Aldrich
Anti-Rac1 Antibody, clone 23A8, clone 23A8, Upstate®, from mouse
Sigma-Aldrich
Rac1/cdc42 Activation Magnetic Beads Pulldown Assay, The Rac1/cdc42 Activation Magnetic Beads Pulldown Assay provides an effective method for detecting Rac & Cdc42 activity in cell lysates with higher yield & easier process utilizing magenetic bead properties.