Merck
  • Phosphorylation of GAPVD1 Is Regulated by the PER Complex and Linked to GAPVD1 Degradation.
Phosphorylation of GAPVD1 Is Regulated by the PER Complex and Linked to GAPVD1 Degradation.
International journal of molecular sciences (2021-05-01)
Hussam Ibrahim, Philipp Reus, Anna Katharina Mundorf, Anna-Lena Grothoff, Valerie Rudenko, Christina Buschhaus, Anja Stefanski, Niklas Berleth, Björn Stork, Kai Stühler, Faiza Kalfalah, Hans Reinke
ABSTRACT
Repressor protein period (PER) complexes play a central role in the molecular oscillator mechanism of the mammalian circadian clock. While the main role of nuclear PER complexes is transcriptional repression, much less is known about the functions of cytoplasmic PER complexes. We found with a biochemical screen for PER2-interacting proteins that the small GTPase regulator GTPase-activating protein and VPS9 domain-containing protein 1 (GAPVD1), which has been identified previously as a component of cytoplasmic PER complexes in mice, is also a bona fide component of human PER complexes. We show that in situ GAPVD1 is closely associated with casein kinase 1 delta (CSNK1D), a kinase that regulates PER2 levels through a phosphoswitch mechanism, and that CSNK1D regulates the phosphorylation of GAPVD1. Moreover, phosphorylation determines the kinetics of GAPVD1 degradation and is controlled by PER2 and a C-terminal autoinhibitory domain in CSNK1D, indicating that the regulation of GAPVD1 phosphorylation is a novel function of cytoplasmic PER complexes and might be part of the oscillator mechanism or an output function of the circadian clock.
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%