Merck
  • A 4-aminoquinoline derivative that markedly sensitizes tumor cell killing by Akt inhibitors with a minimum cytotoxicity to non-cancer cells.
A 4-aminoquinoline derivative that markedly sensitizes tumor cell killing by Akt inhibitors with a minimum cytotoxicity to non-cancer cells.
European journal of medicinal chemistry (2009-12-01)
Changkun Hu, V Raja Solomon, Pablo Cano, Hoyun Lee
ABSTRACT
The purpose of this study was to evaluate the enhancement value of chloroquine analogs when used in combination with Akt inhibitors on the MDA-MB468, MDA-MB231 and MCF7 human breast cancer cell lines. The result showed that the combination of certain chloroquine analogs and Akt inhibitors are highly effective. In particular, the chloroquine analog N'-(7-fluoro-quinolin-4-yl)-N,N-dimethyl-ethane-1,2-diamine (compound 5) was highly effective in sensitizing cancer cell killing when combined with either Akt inhibitor 8 (1-{1-[4-(7-phenyl-1H-imidazo[4,5-g]quinoxalin-6-yl)-benzyl]-piperidin-4-yl}-1,3-dihydro-benzoimidazol-2-one) or 9 ([4-(2-chloro-4a,10a-dihydro-phenoxazin-10-yl)-butyl]-diethyl-amine hydrochloride). Importantly, the enhancement of chloroquine analogs 5 on cell killing by Akt inhibitors 8 and 9 was cancer-specific. Thus, this combinational approach is highly promising in controlling tumors with a minimum side effect. Structural analysis of effective and ineffective chloroquine analogs suggests that the 4-aminoquinoline scaffold and lateral side chain of dimethylamino functionality play an important role for the enhancement of cell killing by Akt inhibitors.
Sigma-Aldrich
Akt Inhibitor X, The Akt Inhibitor X, also referenced under CAS 925681-41-0, controls the biological activity of Akt. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
Akt Inhibitor IV, The Akt Inhibitor IV, also referenced under CAS 681281-88-9, controls the biological activity of Akt. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
Akt Inhibitor VIII, Isozyme-Selective, Akti-1/2, Akt Inhibitor VIII, Isozyme-Selective, Akti-1/2, CAS 612847-09-3, is a cell-permeable, reversible inhibitor of Akt1/Akt2 (IC₅₀ = 58 nM, 210 nM, & 2.12 µM for Akt1, Akt2, and Akt3, respectively).
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)