Merck
  • Distinct Molecular Profiles and Immunotherapy Treatment Outcomes of V600E and V600K BRAF-Mutant Melanoma.
Distinct Molecular Profiles and Immunotherapy Treatment Outcomes of V600E and V600K BRAF-Mutant Melanoma.
Clinical cancer research : an official journal of the American Association for Cancer Research (2019-01-12)
Inês Pires da Silva, Kevin Y X Wang, James S Wilmott, Jeff Holst, Matteo S Carlino, John J Park, Camelia Quek, Matthew Wongchenko, Yibing Yan, Graham Mann, Douglas B Johnson, Jennifer L McQuade, Rajat Rai, Richard F Kefford, Helen Rizos, Richard A Scolyer, Jean Y H Yang, Georgina V Long, Alexander M Menzies
ABSTRACT
BRAF V600E and V600K melanomas have distinct clinicopathologic features, and V600K appear to be less responsive to BRAFi±MEKi. We investigated mechanisms for this and explored whether genotype affects response to immunotherapy. Pretreatment formalin-fixed paraffin-embedded tumors from patients treated with BRAFi±MEKi underwent gene expression profiling and DNA sequencing. Molecular results were validated using The Cancer Genome Atlas (TCGA) data. An independent cohort of V600E/K patients treated with anti-PD-1 immunotherapy was examined. Baseline tissue and clinical outcome with BRAFi±MEKi were studied in 93 patients (78 V600E, 15 V600K). V600K patients had numerically less tumor regression (median, -31% vs. -52%, P = 0.154) and shorter progression-free survival (PFS; median, 5.7 vs. 7.1 months, P = 0.15) compared with V600E. V600K melanomas had lower expression of the ERK pathway feedback regulator dual-specificity phosphatase 6, confirmed with TCGA data (116 V600E, 17 V600K). Pathway analysis showed V600K had lower expression of ERK and higher expression of PI3K-AKT genes than V600E. Higher mutational load was observed in V600K, with a higher proportion of mutations in PIK3R1 and tumor-suppressor genes. In patients treated with anti-PD-1, V600K (n = 19) had superior outcomes than V600E (n = 84), including response rate (53% vs. 29%, P = 0.059), PFS (median, 19 vs. 2.7 months, P = 0.049), and overall survival (20.4 vs. 11.7 months, P = 0.081). BRAF V600K melanomas appear to benefit less from BRAFi±MEKi than V600E, potentially due to less reliance on ERK pathway activation and greater use of alternative pathways. In contrast, these melanomas have higher mutational load and respond better to immunotherapy.