Merck
  • STAT3 is activated in multicellular spheroids of colon carcinoma cells and mediates expression of IRF9 and interferon stimulated genes.
STAT3 is activated in multicellular spheroids of colon carcinoma cells and mediates expression of IRF9 and interferon stimulated genes.
Scientific reports (2019-01-27)
Elin Edsbäcker, Jason T Serviss, Iryna Kolosenko, Caroline Palm-Apergi, Angelo De Milito, Katja Pokrovskaja Tamm
ABSTRACT
Three-dimensional cell cultures, such as multicellular spheroids (MCS), reflect the in vivo architecture of solid tumours and multicellular drug resistance. We previously identified interferon regulatory factor 9 (IRF9) to be responsible for the up-regulation of a subset of interferon (IFN)-stimulated genes (ISGs) in MCS of colon carcinoma cells. This set of ISGs closely resembled a previously identified IFN-related DNA-damage resistance signature (IRDS) that was correlated to resistance to chemo- and radiotherapy. In this study we found that transcription factor STAT3 is activated upstream of IRF9 and binds to the IRF9 promoter in MCS of HCT116 colorectal carcinoma cells. Transferring conditioned media (CM) from high cell density conditions to non-confluent cells resulted in STAT3 activation and increased expression of IRF9 and a panel of IRDS genes, also observed in MCS, suggesting the involvement of a soluble factor. Furthermore, we identified gp130/JAK signalling to be responsible for STAT3 activation, IRF9, and IRDS gene expression in MCS and by CM. Our data suggests a novel mechanism where STAT3 is activated in high cell density conditions resulting in increased expression of IRF9 and, in turn, IRDS genes, underlining a mechanism by which drug resistance is regulated.
Sigma-Aldrich
IgG from goat serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder