Merck
  • Transcriptional repression of IKKβ by p53 in arsenite-induced GADD45α accumulation and apoptosis.
Transcriptional repression of IKKβ by p53 in arsenite-induced GADD45α accumulation and apoptosis.
Oncogene (2018-09-05)
Yongliang Hu, Rui Jin, Ming Gao, Huan Xu, Shuxian Zou, Xiaoguang Li, Chen Xing, Qiyu Wang, Hongli Wang, Jiannan Feng, Meiru Hu, Lun Song
ABSTRACT
Our previous studies revealed that GADD45α is a liable protein, which undergoes MDM2-dependent constitutive ubiquitination and degradation in resting HepG2 hepatoma cells. Arsenite exposure induces ribosomal stress responses mediated by the ribosomal protein S7, which can block MDM2 activity and result in GADD45α accumulation and cell apoptosis. In the present study, we found that one of the catalytic subunits of IκB kinase (IKK), IKKβ, exerted a novel IKKα- and NF-κB-independent function in stabilizing MDM2 and therefore contributed to ubiquitination-dependent degradation of GADD45α in resting HepG2 cells. Arsenite stimulation induced transactivation of p53, which formed a complex with its downstream target, Ets-1, and then synergistically repressed IKKβ transcription, reduced MDM2 stability, and ultimately removed the inhibitory effect of MDM2 on GADD45α induction. In addition, DAPK1 functioned as an upstream protein kinase triggering p53/Ets-1-dependent IKKβ and MDM2 reduction and GADD45α accumulation, thus promoting apoptosis in HepG2 cells. Subsequent studies further revealed that the activation of the DAPK1/p53/Ets-1/IKKβ/MDM2/GADD45α cascade was a common signaling event in mediating apoptosis of diverse cancer cells induced by arsenite and other tumor therapeutic agents. Therefore, we conclude that data in the current study have revealed a novel role for IKKβ in negatively regulating GADD45α protein stability and the contribution of p53-dependent IKKβ reduction to mediating cancer cell apoptosis.
Sigma-Aldrich
Anti-MDM2 (Ab-1) Mouse mAb (IF2), liquid, clone IF2, Calbiochem®
Pricing and availability is not currently available.