Merck
  • Developmental nicotine exposure elicits multigenerational disequilibria in proBDNF proteolysis and glucocorticoid signaling in the frontal cortices, striata, and hippocampi of adolescent mice.
Developmental nicotine exposure elicits multigenerational disequilibria in proBDNF proteolysis and glucocorticoid signaling in the frontal cortices, striata, and hippocampi of adolescent mice.
Biochemical pharmacology (2019-08-14)
Jordan M Buck, Heidi C O'Neill, Jerry A Stitzel
ABSTRACT
Maternal smoking of conventional or vapor cigarettes during pregnancy, a form of developmental nicotine exposure (DNE), enhances the risk of neurodevelopmental disorders such as ADHD, autism, and schizophrenia in children. Modeling the multigenerational effects of smoking during pregnancy and nursing in the first- (F1) and second- (F2) generation adolescent offspring of oral nicotine-treated female C57BL/6J mice, we have previously reported that DNE precipitates intergenerational transmission of nicotine preference, hyperactivity and impulsivity-like behaviors, altered rhythmicity of home cage activity, corticostriatal nicotinic acetylcholine receptor and dopamine transporter dysfunction, and corticostriatal global DNA methylome deficits. In aggregate, these DNE-evoked behavioral, neuropharmacological, and epigenomic anomalies mirror fundamental etiological aspects of neurodevelopmental disorders including ADHD, autism, and schizophrenia. Expanding this line of research, the current study profiled the multigenerational neurotrophic and neuroendocrine consequences of DNE. Results reveal impaired proBDNF proteolysis as indicated by proBDNF-BDNF imbalance, downregulation of the proBDNF processing enzyme furin, atypical glucocorticoid receptor (GR) activity as implied by decreased relative nuclear GR localization, and deficient basal plasma corticosterone (CORT) levels in adolescent DNE offspring and grandoffspring. Collectively, these data recapitulate the BDNF deficits and HPA axis dysregulation characteristic of neurodevelopmental disorders such as ADHD, autism, and schizophrenia as well as the children of maternal smokers. Notably, as BDNF is a quintessential mediator of neurodevelopment, our prior findings of multigenerational DNE-induced behavioral and neuropharmacological abnormalities may stem from neurodevelopmental insults conferred by the proBDNF-BDNF imbalance detected in DNE mice. Similarly, our findings of multigenerational GR hypoactivity may contribute to the increased risk-taking behaviors and aberrant circadian rhythmicity of home cage activity that we previously documented in first- and second-generation DNE mice.
Sigma-Aldrich
Anti-phospho Glucocorticoid Receptor Antibody (Ser287), from rabbit, purified by affinity chromatography