Merck
  • Small Molecule Inhibitors of DYRK1A Identified by Computational and Experimental Approaches.
Small Molecule Inhibitors of DYRK1A Identified by Computational and Experimental Approaches.
International journal of molecular sciences (2020-09-23)
Hye Ree Yoon, Anand Balupuri, Kwang-Eun Choi, Nam Sook Kang
ABSTRACT
Dual-specificity tyrosine phosphorylation-regulated kinase 1A (DYRK1A) is a protein kinase with diverse functions in cell regulation. Abnormal expression and activity of DYRK1A contribute to numerous human malignancies, Down syndrome, and Alzheimer's disease. Notably, DYRK1A has been proposed as a potential therapeutic target for the treatment of diabetes because of its key role in pancreatic β-cell proliferation. Consequently, DYRK1A is an attractive drug target for a variety of diseases. Here, we report the identification of several DYRK1A inhibitors using our in-house topological water network-based approach. All inhibitors were further verified by in vitro assay.
Sigma-Aldrich
SB 415286, ≥98% (HPLC)