Merck
  • Establishment of a Duchenne muscular dystrophy patient-derived induced pluripotent stem cell line carrying a deletion of exons 51-53 of the dystrophin gene (CCMi003-A).
Establishment of a Duchenne muscular dystrophy patient-derived induced pluripotent stem cell line carrying a deletion of exons 51-53 of the dystrophin gene (CCMi003-A).
Stem cell research (2019-08-30)
Davide Rovina, Elisa Castiglioni, Andrea Farini, Marzia Bellichi, Cristina Gervasini, Stefania Paganini, Marina Di Segni, Rosaria Santoro, Yvan Torrente, Giulio Pompilio, Aoife Gowran
ABSTRACT
Duchenne's muscular dystrophy (DMD) is a neuromuscular disorder affecting skeletal and cardiac muscle function, caused by mutations in the dystrophin (DMD) gene. Dermal fibroblasts, isolated from a DMD patient with a reported deletion of exons 51 to 53 in the DMD gene, were reprogramed into induced pluripotent stem cells (iPSCs) by electroporation with episomal vectors containing the reprograming factors: OCT4, SOX2, LIN28, KLF4, and L-MYC. The obtained iPSC line showed iPSC morphology, expression of pluripotency markers, possessed trilineage differentiation potential and was karyotypically normal.