Merck
  • Brazilian red propolis extract enhances expression of antioxidant enzyme genes in vitro and in vivo.
Brazilian red propolis extract enhances expression of antioxidant enzyme genes in vitro and in vivo.
Bioscience, biotechnology, and biochemistry (2020-06-04)
Sho Hotta, Satoshi Uchiyama, Kenji Ichihara
ABSTRACT
Brazilian red propolis reportedly has reactive oxygen species (ROS) scavenging effects in vitro, but the cellular mechanisms remain unclear. In the present study, the effects of an ethanol extract of Brazilian red propolis (EERP) on the Nrf2-ARE intracellular antioxidant pathway were examined in vitro and in vivo. EERP and its constituents transactivated the reporter gene through the ARE sequence and enhanced the expression of Nrf2-regulated genes in HEK293 cells. It also increased Nrf2 protein in the nucleus, which was partially inhibited by kinase inhibitors. Furthermore, EERP suppressed ROS generation and cytotoxicity induced by tert-butyl hydroperoxide. In vivo, orally administered EERP increased the expression of Nrf2-regulated genes in mice liver. These results suggest that EERP is a potential resource for preventing oxidative stress-related diseases as an Nrf2 inducer.
Sigma-Aldrich
4-Bromoanisole, ≥99.0%
Sigma-Aldrich
SL327, ≥98% (HPLC), Mixture of E & Z isomers, solid
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
SP600125, ≥98% (HPLC)