Merck
  • Exploring the Lower Limit of Individual DNA-Encoded Library Molecules in Selection.
Exploring the Lower Limit of Individual DNA-Encoded Library Molecules in Selection.
SLAS discovery : advancing life sciences R & D (2019-12-21)
Qiuxia Chen, Xuemin Cheng, Lifang Zhang, Xianyang Li, Purui Chen, Jian Liu, Lanjun Zhang, Hong Wei, Zhonghan Li, Dengfeng Dou
ABSTRACT
DNA-encoded library (DEL) technology has been used as an ultra-high-throughput screening approach for hit identification of drug targets. This process is an affinity-based selection and requires incubation of DEL molecules with the target. Currently, in most reported cases, the input (i.e., the copy number) of individual DEL molecules varies from 105 to 107. With the ever-increasing DEL size and screening cost, lowering the input of DEL molecules while maintaining an appropriate signal-to-noise ratio in a selection is of paramount importance. In this article, we varied the input of DEL ranging from 103 to 105 in selections with two different protein targets to explore the lower limit of DEL molecule input. The results could facilitate the optimization of the DEL selection process and reduce costs related to library consumption.
Sigma-Aldrich
Deoxyribonucleic acid, low molecular weight from salmon sperm