Merck
  • Influence of the particle size of gadolinium-loaded chitosan nanoparticles on their tumor-killing effect in neutron capture therapy in vitro.
Influence of the particle size of gadolinium-loaded chitosan nanoparticles on their tumor-killing effect in neutron capture therapy in vitro.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2020-08-21)
Tooru Andoh, Yugo Nakatani, Minoru Suzuki, Yoshinori Sakurai, Takuya Fujimoto, Hideki Ichikawa
ABSTRACT
Neutron capture therapy using 157Gd (Gd-NCT) is currently under development as a cancer radiotherapy. Melanoma cells were treated with gadolinium-loaded chitosan nanoparticles (Gd-nanoCPs) for Gd-NCT. Smaller Gd-nanoCPs had higher Gd content and better cellular association of Gd and thereby made the tumor-killing effect more efficient in comparison to larger Gd-nanoCPs. This indicates that Gd-nanoCP size reduction is an efficient method for improving the cellular affinity of Gd-nanoCPs and for enhancing the tumor-killing effect of Gd-NCT.
Sigma-Aldrich
Diethylenetriaminepentaacetic acid gadolinium(III) dihydrogen salt hydrate, 97%