Merck
  • A Fe3+-crosslinked pyrogallol-tethered gelatin adhesive hydrogel with antibacterial activity for wound healing.
A Fe3+-crosslinked pyrogallol-tethered gelatin adhesive hydrogel with antibacterial activity for wound healing.
Biomaterials science (2020-05-05)
Ning Han, Ziyang Xu, Chunyan Cui, Yuan Li, Dongfei Zhang, Meng Xiao, Chuanchuan Fan, Tengling Wu, Jianhai Yang, Wenguang Liu
ABSTRACT
In this work, a tunicate-inspired gelatin-based hydrogel is prepared by simply mixing 2,3,4-trihydroxybenzaldehyde (THB)-tethered gelatin solution with a small amount of Fe3+ ions via the Schiff-base reaction and simultaneous formation of hexavalent Fe-complexes. The resulting hydrogel (termed GelTHB-Fe) exhibits not only tunable gelation time, rheological properties and self-healing ability by adjusting the composition, but also robust adhesion to a variety of materials, with an average adhesion strength of 136.7 kPa, 147.3 kPa, 153.7 kPa, 92.9 kPa, and 56.5 kPa to PMMA, iron, ceramics, glass and pigskin, respectively. Intriguingly, the pyrogallol moieties impart an antibacterial activity to the GelTHB-Fe hydrogel, which is shown to reduce infection and promote wound healing in a diabetic rat model. This GelTHB-Fe hydrogel holds great potential as a promising tissue adhesive.
Sigma-Aldrich
2,3,4-Trihydroxybenzaldehyde, 98%