Merck
  • Efficacy trials in turkey poults with tylosin tartrate against Mycoplasma gallisepticum and Mycoplasma meleagridis.
Efficacy trials in turkey poults with tylosin tartrate against Mycoplasma gallisepticum and Mycoplasma meleagridis.
Research in veterinary science (1975-11-01)
D R Wise, M K Fuller
ABSTRACT
Tylosin tartrate, administered in the drinking water at a concentration of 0.55 g/litre for the first three days after hatching, was highly effective in controlling the adverse consequences of a Mycoplasma gallisepticum infection, established by air sac injection at one day of age, in turkey poults. Tylosin was ineffective in controlling M meleagridis infections established in embryo or at one day of age when administered in the drinking water of poults. Both mycoplasma isolates used were inhibited in vitro by a tylosin concentration of 0.1 mug/ml.
Sigma-Aldrich
Tylosin tartrate, potency: ≥800 units/mg tylosin