Merck
  • Comprehensive analysis of posttranslational protein modifications in aging of subcellular compartments.
Comprehensive analysis of posttranslational protein modifications in aging of subcellular compartments.
Scientific reports (2020-05-07)
Tim Baldensperger, Michael Eggen, Jonas Kappen, Patrick R Winterhalter, Thorsten Pfirrmann, Marcus A Glomb
ABSTRACT
Enzymatic and non-enzymatic posttranslational protein modifications by oxidation, glycation and acylation are key regulatory mechanisms in hallmarks of aging like inflammation, altered epigenetics and decline in proteostasis. In this study a mouse cohort was used to monitor changes of posttranslational modifications in the aging process. A protocol for the extraction of histones, cytosolic and mitochondrial proteins from mouse liver was developed and validated. In total, 6 lysine acylation structures, 7 advanced glycation endproducts, 6 oxidative stress markers, and citrullination were quantitated in proteins of subcellular compartments using HPLC-MS/MS. Methionine sulfoxide, acetylation, formylation, and citrullination were the most abundant modifications. Histone proteins were extraordinary high modified and non-enzymatic modifications accumulated in all subcellular compartments during the aging process. Compared to acetylation of histone proteins which gave between 350 and 305 µmol/mol leucine equivalents in young and old animals, modifications like acylation, glycation, and citrullination raised to 43%, 20%, and 18% of acetylation, respectively. On the other hand there was an age related increase of selected oxidative stress markers by up to 150%. The data and patterns measured in this study are mandatory for further studies and will strongly facilitate understanding of the molecular mechanisms in aging.
Sigma-Aldrich
Nε-Acetyl-L-lysine
Sigma-Aldrich
L-Methionine sulfoxide
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid