Merck
  • Structure and thermal stability of pyruvated carrageenans from the red alga Coccotylus truncatus.
Structure and thermal stability of pyruvated carrageenans from the red alga Coccotylus truncatus.
Carbohydrate research (2009-03-10)
Rando Tuvikene, Kalle Truus, Marju Robal, Tõnis Pehk, Tiiu Kailas, Merike Vaher, Tiina Paalme
ABSTRACT
The composition, structure, and thermal stability of carrageenans from unattached Coccotylus truncatus (the Baltic Sea, Estonia) were investigated. The complex polysaccharide was characterized by (13)C NMR and FTIR spectroscopy, ICP-OES and gel permeation chromatography methods. The main components of C. truncatus galactan are 3,6-anhydro-alpha-D-galactose-2-sulfate (30+/-1.5%) and beta-D-galactose-4-sulfate (45.3%), indicating a iota-carrageenan backbone. As the minor components, alpha-D-galactose-2,6-disulfate (12+/-2%) from nu-carrageenan and 4',6'-pyruvated beta-D-galactopyranosyl residues (1.4%) from pyruvated alpha-carrageenan are found to be present, latter being responsible for the undersulfated nature of the galactan. The native polysaccharide with the average molecular weight of about 1500 kDa is highly susceptible to thermal degradation. The high-temperature treatment of this galactan gives products with 3,6-anhydro-alpha-D-galactose units predominantly at the reducing end. The carrageenan extraction from C. truncatus gives characteristically low yields (12-17%); weak gelling ability of the polysaccharides from this seaweed species (gel strength 30-40 g/cm(2)) does not depend significantly on extraction conditions.