Merck
  • Early alteration of epigenetic-related transcription in Huntington's disease mouse models.
Early alteration of epigenetic-related transcription in Huntington's disease mouse models.
Scientific reports (2018-07-04)
Irati Hervás-Corpión, Deisy Guiretti, Manuel Alcaraz-Iborra, Román Olivares, Antonio Campos-Caro, Ángel Barco, Luis M Valor
ABSTRACT
Transcriptional dysregulation in Huntington's disease (HD) affects the expression of genes involved in survival and neuronal functions throughout the progression of the pathology. In recent years, extensive research has focused on epigenetic and chromatin-modifying factors as a causative explanation for such dysregulation, offering attractive targets for pharmacological therapies. In this work, we extensively examined the gene expression profiles in the cortex, striatum, hippocampus and cerebellum of juvenile R6/1 and N171-82Q mice, models of rapidly progressive HD, to retrieve the early transcriptional signatures associated with this pathology. These profiles were largely consistent across HD datasets, contained tissular and neuronal-specific genes and showed significant correspondence with the transcriptional changes in mouse strains deficient for epigenetic regulatory genes. The most prominent cases were the conditional knockout of the lysine acetyltransferase CBP in post-mitotic forebrain neurons, the double knockout of the histone methyltransferases Ezh1 and Ezh2, components of the polycomb repressor complex 2 (PRC2), and the conditional mutants of the histone methyltransferases G9a (Ehmt2) and GLP (Ehmt1). Based on these observations, we propose that the neuronal epigenetic status is compromised in the prodromal stages of HD, leading to an altered transcriptional programme that is prominently involved in neuronal identity.
Sigma-Aldrich
Anti-MMP-1 Antibody, hinge region, from rabbit, purified by affinity chromatography