Merck
  • A miRNA signature in endothelial cell-derived extracellular vesicles in tumor-bearing mice.
A miRNA signature in endothelial cell-derived extracellular vesicles in tumor-bearing mice.
Scientific reports (2019-11-16)
James V McCann, Amber Liu, Luca Musante, Uta Erdbrügger, Joanne Lannigan, Andrew C Dudley
ABSTRACT
Extracellular vesicles (EVs) play important roles in tumor progression by altering immune surveillance, promoting vascular dysfunction, and priming distant sites for organotropic metastases. The miRNA expression patterns in circulating EVs are important diagnostic tools in cancer. However, multiple cell types within the tumor microenvironment (TME) including cancer cells and stromal cells (e.g. immune cells, fibroblasts, and endothelial cells, ECs) contribute to the pool of circulating EVs. Because EVs of different cellular origins have different functional properties, auditing the cargo derived from cell type-specific EVs in the TME is essential. Here, we demonstrate that a murine EC lineage-tracing model (Cdh5-CreERT2:ZSGreenl/s/l mice) can be used to isolate EC-derived extracellular vesicles (EC-EVs). We further show that purified ZSGreen+ EVs express expected EV markers, they are transferable to multiple recipient cells, and circulating EC-EVs from tumor-bearing mice harbor elevated levels of specific miRNAs (e.g. miR-30c, miR-126, miR-146a, and miR-125b) compared to non tumor-bearing counterparts. These results suggest that, in the tumor setting, ECs may systemically direct the function of heterotypic cell types either in the circulation or in different organ micro-environments via the cargo contained within their EVs.
Sigma-Aldrich
Anti-TSG101 antibody produced in rabbit, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution