Merck
  • Distinct Cell-Cycle Control in Two Different States of Mouse Pluripotency.
Distinct Cell-Cycle Control in Two Different States of Mouse Pluripotency.
Cell stem cell (2017-10-07)
Menno Ter Huurne, James Chappell, Stephen Dalton, Hendrik G Stunnenberg
ABSTRACT
Mouse embryonic stem cells (ESCs) cultured in serum are characterized by hyper-phosphorylated RB protein, lack of G1 control, and rapid progression through the cell cycle. Here, we show that ESCs grown in the presence of two small-molecule inhibitors (2i ESCs) have a longer G1-phase with hypo-phosphorylated RB, implying that they have a functional G1 checkpoint. Deletion of RB, P107, and P130 in 2i ESCs results in a G1-phase similar to that of serum ESCs. Inhibition of the ERK signaling pathway in serum ESCs results in the appearance of hypo-phosphorylated RB and the reinstatement of a G1 checkpoint. In addition, induction of a dormant state by the inhibition of MYC, resembling diapause, requires the presence of the RB family proteins. Collectively, our data show that RB-dependent G1 restriction point signaling is active in mouse ESCs grown in 2i but abrogated in serum by ERK-dependent phosphorylation.
Sigma-Aldrich
Anti-E2F-1 Antibody, clones KH20 and KH95, Upstate®, from mouse