Merck
  • Efficient Construction and Effective Screening of Synthetic Domain Antibody Libraries.
Efficient Construction and Effective Screening of Synthetic Domain Antibody Libraries.
Methods and protocols (2019-06-06)
Arghavan Solemani Zadeh, Alissa Grässer, Heiko Dinter, Maximilian Hermes, Katharina Schindowski
ABSTRACT
Phage display is a powerful technique for drug discovery in biomedical research in particular for antibody libraries. But, several technical challenges are associated with the selection process. For instance, during the panning step, the successful elution of the phages bound to the antigen is critical in order to avoid losing the most promising binders. Here, we present an efficient protocol to establish, screen and select synthetic libraries of domain antibodies using phage display. We do not only present suitable solutions to the above-mentioned challenges to improve elution by 50-fold, but we also present a step by step in-depth protocol with miniaturized volumes and optimized procedures to save material, costs and time for a successful phage display with domain antibodies. Hence, this protocol improves the selection process for an efficient handling process. The here presented library is based on the variable domain (vNAR) of the naturally occurring novel antibody receptor (IgNAR) from cartilage fishes. Diversity was introduced in the Complementarity-Determining Region 3 (CDR3) of the antigen-binding site with different composition and length.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Kanamycin sulfate, mixture of Kanamycin A (main component) and Kanamycin B and C
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody