Merck
  • Polycomb group proteins EZH2 and EED directly regulate androgen receptor in advanced prostate cancer.
Polycomb group proteins EZH2 and EED directly regulate androgen receptor in advanced prostate cancer.
International journal of cancer (2019-01-11)
Qipeng Liu, Guangyu Wang, Qiaqia Li, Weihua Jiang, Jung-Sun Kim, Rui Wang, Sen Zhu, Xiaoju Wang, Lin Yan, Yang Yi, Lili Zhang, Qingshu Meng, Chao Li, Dongyu Zhao, Yuanyuan Qiao, Yong Li, Demirkan B Gursel, Arul M Chinnaiyan, Kaifu Chen, Qi Cao
ABSTRACT
Polycomb group proteins are important epigenetic regulators for cell proliferation and differentiation, organ development, as well as initiation and progression of lethal diseases, including cancer. Upregulated Polycomb group proteins, including Enhancer of zeste homolog 2 (EZH2), promote proliferation, migration, invasion and metastasis of cancer cells, as well as self-renewal of cancer stem cells. In our study, we report that EZH2 and embryonic ectoderm development (EED) indicate respective direct interaction with androgen receptor (AR). In the context of AR-positive prostate cancer, EZH2 and EED regulate AR expression levels and AR downstream targets. More importantly, we demonstrate that targeting EZH2 with the small-molecule inhibitor astemizole in cancer significantly represses the EZH2 and AR expression as well as the neoplastic capacities. These results collectively suggest that pharmacologically targeting EZH2 might be a promising strategy for advanced prostate cancer.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Normal Mouse IgG, Normal Mouse IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.
Sigma-Aldrich
Normal Rabbit IgG, Normal Rabbit IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.
Sigma-Aldrich
Anti-EED Antibody, from rabbit
Sigma-Aldrich
Anti-Androgen Receptor Antibody, from rabbit, purified by affinity chromatography