Merck
  • Endogenous peptide LYENRL prevents the activation of hypertrophic scar-derived fibroblasts by inhibiting the TGF-β1/Smad pathway.
Endogenous peptide LYENRL prevents the activation of hypertrophic scar-derived fibroblasts by inhibiting the TGF-β1/Smad pathway.
Life sciences (2019-07-26)
Xiaojun Ji, Zhe Tang, Weiwei Shuai, Zhirui Zhang, Jingyun Li, Ling Chen, Jing Cao, Wu Yin
ABSTRACT
Hypertrophic scar formation is a fibroproliferative disorder caused by abnormal wound healing. At present, there are limited treatment strategies for hypertrophic scars. In this study, we identified an endogenous peptide, LYENRL, through peptidomics screening that is downregulated in scar skin tissues. The peptide exhibited concentration dependent inhibitory effects on the proliferation, migration and extracellular matrix (ECM) production of scar fibroblasts. By eukaryotic transcriptome sequencing analysis, we noted that LYENRL downregulated gene sets in scar fibroblasts were associated with the transforming growth factor-β (TGF-β) signaling pathway. Further experiments revealed that LYENRL was able to inhibit the activation of TGF-β1/Smad signaling and TGF-β1-induced activation of scar fibroblasts at the source by blocking the binding of AP-1 to the corresponding region of the Tgfb1 promoter, which in turn inhibited gene expression of Tgfb1. Taken together, we concluded that the effects of LYENRL on scar fibroblasts make it a potential peptide drug for hypertrophic scar treatment.